Assay Details
| Gene Symbol: RPL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014894.1 | NP_001014894.1 | 9 - 10 | 1082 | 65 | |
| GenBank mRNA | BC102521.1 | - | 9 - 10 | 1067 | 65 | |
| BT021487.1 | - | 9 - 10 | 1082 | 65 | ||
More Information
Set Membership: |
|

RPL4
Genomic Map