Assay Details
| Gene Symbol: MAPK1IP1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014963.2 | NP_001014963.1 | 1 - 2 | 180 | 69 | |
| GenBank mRNA | BC118396.1 | - | 1 - 2 | 152 | 69 | |
| BT020907.1 | - | 149 | 69 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC118396.1, NM_001014963.2 transcripts.
Set Membership: |
|

MAPK1IP1L
Genomic Map