Assay Details
| Gene Symbol: OSER1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015568.1 | NP_001015568.1 | 1 - 2 | 68 | 99 | |
| GenBank mRNA | BC110226.1 | - | 1 - 2 | 129 | 99 | |
| BT021020.1 | - | 1 - 2 | 68 | 99 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC110226.1, NM_001015568.1 transcripts.
Set Membership: |
|

OSER1
Genomic Map