Assay Details
| Gene Symbol: ACKR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015581.1 | NP_001015581.1 | 1 - 2 | 40 | 118 | |
| GenBank mRNA | BT021494.1 | - | 1 - 2 | 40 | 118 | |
| BT021720.1 | - | 1 - 2 | 57 | 118 | ||
| BT021828.1 | - | 1 - 2 | 40 | 118 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001015581.1 transcripts.
Set Membership: |
|

ACKR2
Genomic Map