Assay Details
| Gene Symbol: WDR77 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015584.1 | NP_001015584.1 | 6 - 7 | 684 | 76 | |
| GenBank mRNA | BC133485.1 | - | 6 - 7 | 686 | 76 | |
| BT020933.1 | - | 6 - 7 | 684 | 76 | ||
More Information
Set Membership: |
|

WDR77
Genomic Map