Assay Details
| Gene Symbol: KRT19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015600.4 | NP_001015600.1 | 4 - 5 | 878 | 70 | |
| GenBank mRNA | BC116088.1 | - | 4 - 5 | 864 | 70 | |
More Information
Set Membership: |
|

KRT19
Genomic Map