Assay Details
| Gene Symbol: CLIC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015608.1 | NP_001015608.1 | 5 - 6 | 732 | 76 | |
| GenBank mRNA | AY999167.1 | - | 2 - 3 | 256 | 76 | |
| BC102103.1 | - | 5 - 6 | 583 | 76 | ||
| BT021003.1 | - | 5 - 6 | 732 | 76 | ||
More Information
Set Membership: |
|

CLIC1
Genomic Map