Assay Details
| Gene Symbol: PSMB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015615.1 | NP_001015615.1 | 3 - 4 | 357 | 59 | |
| GenBank mRNA | BC102367.1 | - | 3 - 4 | 359 | 59 | |
| BT020855.1 | - | 3 - 4 | 332 | 59 | ||
| BT020920.1 | - | 3 - 4 | 357 | 59 | ||
More Information
Set Membership: |
|

PSMB2
Genomic Map