Assay Details
| Gene Symbol: CFL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015655.1 | NP_001015655.1 | 1 - 2 | 74 | 101 | |
| GenBank mRNA | BC103077.1 | - | 1 - 2 | 84 | 101 | |
| BT020963.1 | - | 1 - 2 | 74 | 101 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC103077.1, NM_001015655.1 transcripts.
Set Membership: |
|

CFL1
Genomic Map