Assay Details
| Gene Symbol: VPS72 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015658.1 | NP_001015658.1 | 5 - 6 | 761 | 73 | |
| GenBank mRNA | BC109639.1 | - | 5 - 6 | 726 | 73 | |
| BT020964.1 | - | 5 - 6 | 761 | 73 | ||
More Information
Set Membership: |
|

VPS72
Genomic Map