Assay Details
| Gene Symbol: ZFYVE27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017939.1 | NP_001017939.1 | 3 - 4 | 430 | 55 | |
| GenBank mRNA | BC134516.1 | - | 3 - 4 | 386 | 55 | |
| BT021199.1 | - | 3 - 4 | 430 | 55 | ||
More Information
Set Membership: |
|

ZFYVE27
Genomic Map