Assay Details
| Gene Symbol: PRMT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024495.1 | NP_001019666.1 | 1 - 2 | 107 | 60 | |
| GenBank mRNA | BC104629.1 | - | 1 - 2 | 79 | 60 | |
| BT021771.1 | - | 2 - 3 | 126 | 60 | ||
| BT021778.1 | - | 1 - 2 | 107 | 60 | ||
More Information
Set Membership: |
|

PRMT2
Genomic Map