Assay Details
| Gene Symbol: SESN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024507.1 | NP_001019678.1 | 3 - 4 | 648 | 63 | |
| GenBank mRNA | BC123386.1 | - | 3 - 4 | 696 | 63 | |
| BT021909.1 | - | 3 - 4 | 648 | 63 | ||
More Information
Set Membership: |
|

SESN2
Genomic Map