Assay Details
| Gene Symbol: CORO1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113243.1 | NP_001106714.1 | 2 - 3 | 273 | 62 | |
| GenBank mRNA | BC103376.1 | - | 2 - 3 | 255 | 62 | |
| BT021806.1 | - | 2 - 3 | 243 | 62 | ||
| DT824953.1 | - | 2 - 3 | 260 | 62 | ||
| EE355537.1 | - | 2 - 3 | 273 | 62 | ||
More Information
Set Membership: |
|

CORO1B
Genomic Map