Assay Details
| Gene Symbol: RAB13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024540.1 | NP_001019711.1 | 1 - 2 | 235 | 103 | |
| GenBank mRNA | BC109840.1 | - | 1 - 2 | 237 | 103 | |
| BT021522.1 | - | 1 - 2 | 235 | 103 | ||
More Information
Set Membership: |
|

RAB13
Genomic Map