Assay Details
| Gene Symbol: EMP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024562.1 | NP_001019733.1 | 1 - 2 | 276 | 84 | |
| GenBank mRNA | BC126787.1 | - | 1 - 2 | 60 | 84 | |
| BT021531.1 | - | 1 - 2 | 276 | 84 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC126787.1, NM_001024562.1 transcripts.
Set Membership: |
|

EMP3
Genomic Map