Assay Details
| Gene Symbol: EMP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024562.1 | NP_001019733.1 | 3 - 4 | 535 | 66 | |
| GenBank mRNA | BC126787.1 | - | 3 - 4 | 319 | 66 | |
| BT021531.1 | - | 3 - 4 | 535 | 66 | ||
More Information
Set Membership: |
|

EMP3
Genomic Map