Assay Details
| Gene Symbol: RPLP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025340.2 | NP_001020511.1 | 2 - 3 | 274 | 72 | |
| GenBank mRNA | AY911313.1 | - | 2 - 3 | 275 | 72 | |
| BC102695.1 | - | 1 | 276 | 72 | ||
More Information
Set Membership: |
|

RPLP1
Genomic Map