Assay Details
| Gene Symbol: IRF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029845.3 | NP_001025016.1 | 4 - 5 | 637 | 76 | |
| GenBank mRNA | AJ879589.1 | - | 3 - 4 | 431 | 76 | |
| BC102119.1 | - | 4 - 5 | 638 | 76 | ||
| EU194377.1 | - | 3 - 4 | 410 | 76 | ||
| KU641390.1 | - | 4 - 5 | 478 | 76 | ||
More Information
Set Membership: |
|

IRF3
Genomic Map