Assay Details
| Gene Symbol: PSMA7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034233.2 | NP_001029405.1 | 2 - 3 | 293 | 74 | |
| GenBank mRNA | AY911343.1 | - | 2 - 3 | 296 | 74 | |
| BC103328.1 | - | 2 - 3 | 294 | 74 | ||
| BC151811.1 | - | 2 - 3 | 311 | 74 | ||
More Information
Set Membership: |
|

PSMA7
Genomic Map