Assay Details
| Gene Symbol: ATF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034342.2 | NP_001029514.1 | 1 - 2 | 196 | 148 | |
| GenBank mRNA | BC102236.1 | - | 1 - 2 | 198 | 148 | |
| BC151812.1 | - | 1 - 2 | 197 | 148 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC102236.1, BC151812.1, NM_001034342.2 transcripts.
Set Membership: |
|

ATF4
Genomic Map