Assay Details
| Gene Symbol: ATF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034342.2 | NP_001029514.1 | 2 - 3 | 505 | 63 | |
| GenBank mRNA | BC102236.1 | - | 2 - 3 | 507 | 63 | |
| BC151812.1 | - | 2 - 3 | 506 | 63 | ||
More Information
Set Membership: |
|

ATF4
Genomic Map