Assay Details
| Gene Symbol: TIMM22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034498.1 | NP_001029670.1 | 3 - 4 | 530 | 66 | |
| GenBank mRNA | BC148116.1 | - | 3 - 4 | 532 | 66 | |
| BT021190.1 | - | 3 - 4 | 530 | 66 | ||
More Information
Set Membership: |
|

TIMM22
Genomic Map