Assay Details
| Gene Symbol: EBP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034500.1 | NP_001029672.1 | 1 - 2 | 44 | 55 | |
| GenBank mRNA | BC103111.1 | - | 1 - 2 | 44 | 55 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC103111.1, NM_001034500.1 transcripts.
Set Membership: |
|

EBP
Genomic Map