Assay Details
| Gene Symbol: TSPAN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034657.1 | NP_001029829.1 | 1 - 2 | 88 | 59 | |
| GenBank mRNA | BT021800.1 | - | 1 - 2 | 88 | 59 | |
More Information
Set Membership: |
|

TSPAN2
Genomic Map