Assay Details
| Gene Symbol: ARPC5L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034737.1 | NP_001029909.1 | 2 - 3 | 298 | 93 | |
| GenBank mRNA | BC120185.1 | - | 2 - 3 | 312 | 93 | |
| BT021057.1 | - | 2 - 3 | 298 | 93 | ||
More Information
Set Membership: |
|

ARPC5L
Genomic Map