Assay Details
| Gene Symbol: F7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034806.1 | NP_001029978.1 | 2 - 3 | 252 | 59 | |
| GenBank mRNA | BC146045.1 | - | 2 - 3 | 267 | 59 | |
| BT021584.1 | - | 2 - 3 | 252 | 59 | ||
More Information
Set Membership: |
|

F7
Genomic Map