Assay Details
| Gene Symbol: FLAD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035039.2 | NP_001030211.2 | 6 - 7 | 1479 | 64 | |
| GenBank mRNA | BC103379.1 | - | 6 - 7 | 1683 | 64 | |
More Information
Set Membership: |
|

FLAD1
Genomic Map