Assay Details
| Gene Symbol: OXNAD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035090.1 | NP_001030262.1 | 5 - 6 | 479 | 59 | |
| GenBank mRNA | BT021570.1 | - | 5 - 6 | 479 | 59 | |
More Information
Set Membership: |
|

OXNAD1
Genomic Map