Assay Details
| Gene Symbol: ALAS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035103.2 | NP_001030275.1 | 10 - 11 | 1649 | 60 | |
| GenBank mRNA | BC102938.1 | - | 10 - 11 | 1650 | 60 | |
More Information
Set Membership: |
|

ALAS2
Genomic Map