Assay Details
| Gene Symbol: ADI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035468.2 | NP_001030545.1 | 3 - 4 | 491 | 55 | |
| GenBank mRNA | BC103235.1 | - | 3 - 4 | 492 | 55 | |
More Information
Set Membership: |
|

ADI1
Genomic Map