Assay Details
| Gene Symbol: HTR4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040485.1 | NP_001035575.1 | 5 - 6 | 598 | 72 | |
| GenBank mRNA | BT020865.1 | - | 5 - 6 | 598 | 72 | |
More Information
Set Membership: |
|

HTR4
Genomic Map