Assay Details
| Gene Symbol: IP6K3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040542.2 | NP_001035632.2 | 4 - 5 | 899 | 55 | |
| GenBank mRNA | BC140544.1 | - | 4 - 5 | 900 | 55 | |
More Information
Set Membership: |
|

IP6K3
Genomic Map