Assay Details
| Gene Symbol: MRPS6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040584.2 | NP_001035674.1 | 2 - 3 | 330 | 91 | |
| GenBank mRNA | BC102978.1 | - | 2 - 3 | 331 | 91 | |
More Information
Set Membership: |
|

MRPS6
Genomic Map