Assay Details
| Gene Symbol: AP1S2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040591.2 | NP_001035681.1 | 2 - 3 | 305 | 83 | |
| GenBank mRNA | BC103139.1 | - | 2 - 3 | 307 | 83 | |
More Information
Set Membership: |
|

AP1S2
Genomic Map