Assay Details
| Gene Symbol: EIF3G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079623.2 | NP_001073091.1 | 2 - 3 | 113 | 54 | |
| GenBank mRNA | BC102984.1 | - | 2 - 3 | 115 | 54 | |
More Information
Set Membership: |
|

EIF3G
Genomic Map