Assay Details
| Gene Symbol: CLYBL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080296.1 | NP_001073765.1 | 3 - 4 | 473 | 60 | |
| GenBank mRNA | BT021750.1 | - | 3 - 4 | 473 | 60 | |
More Information
Set Membership: |
|

CLYBL
Genomic Map