Assay Details
| Gene Symbol: MRPL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080317.2 | NP_001073786.1 | 6 - 7 | 751 | 74 | |
| GenBank mRNA | BC103047.1 | - | 6 - 7 | 752 | 74 | |
More Information
Set Membership: |
|

MRPL3
Genomic Map