Assay Details
| Gene Symbol: DPP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174039.3 | NP_776464.1 | 13 - 14 | 1495 | 83 | |
| GenBank mRNA | AF461806.1 | - | 13 - 14 | 1496 | 83 | |
| AY056834.1 | - | 12 - 13 | 1168 | 83 | ||
| BC102523.1 | - | 13 - 14 | 1261 | 83 | ||
More Information
Set Membership: |
|

DPP4
Genomic Map