Assay Details
| Gene Symbol: DPP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174039.3 | NP_776464.1 | 1 - 2 | 329 | 71 | |
| GenBank mRNA | AF461806.1 | - | 1 - 2 | 330 | 71 | |
| BC102523.1 | - | 1 - 2 | 95 | 71 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC102523.1, NM_174039.3 transcripts.
Set Membership: |
|

DPP4
Genomic Map