Assay Details
| Gene Symbol: DPP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174039.3 | NP_776464.1 | 22 - 23 | 2312 | 63 | |
| GenBank mRNA | AF461806.1 | - | 22 - 23 | 2313 | 63 | |
| AY056834.1 | - | 21 - 22 | 1985 | 63 | ||
| BC102523.1 | - | 22 - 23 | 2078 | 63 | ||
More Information
Set Membership: |
|

DPP4
Genomic Map