Assay Details
| Gene Symbol: DPP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174039.3 | NP_776464.1 | 25 - 26 | 2516 | 66 | |
| GenBank mRNA | AF461806.1 | - | 25 - 26 | 2517 | 66 | |
| AY056834.1 | - | 24 - 25 | 2189 | 66 | ||
| BC102523.1 | - | 25 - 26 | 2282 | 66 | ||
More Information
Set Membership: |
|

DPP4
Genomic Map