Assay Details
| Gene Symbol: DPP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174039.3 | NP_776464.1 | 9 - 10 | 1105 | 97 | |
| GenBank mRNA | AF461806.1 | - | 9 - 10 | 1106 | 97 | |
| AY056834.1 | - | 8 - 9 | 778 | 97 | ||
| BC102523.1 | - | 9 - 10 | 871 | 97 | ||
More Information
Set Membership: |
|

DPP4
Genomic Map