Assay Details
| Gene Symbol: FATE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174052.2 | NP_776477.1 | 3 - 4 | 377 | 66 | |
| GenBank mRNA | AF387304.1 | - | 3 - 4 | 377 | 66 | |
| BC109644.1 | - | 3 - 4 | 395 | 66 | ||
More Information
Set Membership: |
|

FATE1
Genomic Map