Assay Details
| Gene Symbol: TPSB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174202.3 | NP_776627.2 | 5 - 6 | 697 | 103 | |
| GenBank mRNA | AF515641.1 | - | 5 - 6 | 673 | 103 | |
| BC149582.1 | - | 5 - 6 | 677 | 103 | ||
More Information
Set Membership: |
|

TPSB2
Genomic Map