Assay Details
| Gene Symbol: EZR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174217.2 | NP_776642.1 | 4 - 5 | 647 | 57 | |
| GenBank mRNA | BC102573.1 | - | 4 - 5 | 647 | 57 | |
| M98498.1 | - | 4 - 5 | 624 | 57 | ||
More Information
Set Membership: |
|

EZR
Genomic Map