Assay Details
| Gene Symbol: ANXA8L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174241.2 | NP_776666.1 | 1 - 2 | 73 | 62 | |
| GenBank mRNA | AF417637.1 | - | 1 - 2 | 73 | 62 | |
| BC113321.1 | - | 1 - 2 | 122 | 62 | ||
| BT021645.1 | - | 1 - 2 | 105 | 62 | ||
| BT021646.1 | - | 1 - 2 | 105 | 62 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC113321.1, NM_174241.2 transcripts.
Set Membership: |
|

ANXA8L1
Genomic Map