Assay Details
| Gene Symbol: ANXA8L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174241.2 | NP_776666.1 | 2 - 3 | 164 | 67 | |
| GenBank mRNA | AF417637.1 | - | 2 - 3 | 164 | 67 | |
| BC113321.1 | - | 2 - 3 | 213 | 67 | ||
| BT021645.1 | - | 2 - 3 | 196 | 67 | ||
| BT021646.1 | - | 2 - 3 | 196 | 67 | ||
More Information
Set Membership: |
|

ANXA8L1
Genomic Map