Assay Details
| Gene Symbol: CABP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174256.3 | NP_776681.1 | 1 - 2 | 435 | 69 | |
| GenBank mRNA | AY048883.1 | - | 1 - 2 | 436 | 69 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CABP4| Gene Symbol: CABP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174256.3 | NP_776681.1 | 1 - 2 | 435 | 69 | |
| GenBank mRNA | AY048883.1 | - | 1 - 2 | 436 | 69 | |
Set Membership: |
|