Assay Details
| Gene Symbol: LOXL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174383.2 | NP_776808.1 | 2 - 3 | 1399 | 69 | |
| GenBank mRNA | AF421185.1 | - | 2 - 3 | 1399 | 69 | |
| BC126600.1 | - | 2 - 3 | 1583 | 69 | ||
More Information
Set Membership: |
|

LOXL1
Genomic Map